Skip to Content
MilliporeSigma
All Photos(1)

Documents

CTAC

90071812, dog thyroid, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CTAC, 90071812

biological source

dog thyroid

growth mode

Adherent

karyotype

2n = 78

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Dog thyroid gland adenocarcinoma

Cell Line Description

The cell line CTAC originated from a 7 year old female boxer with thyroid adenocarcinoma of small-cell type. CTAC has been reported to be highly susceptible for use in canine natural killer cell (chromium release) assays.

Application

Canine natural killer cell assay.

Culture Medium

RPMI 1640 or MEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service