Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

295337

Sigma-Aldrich

Trifluoromethane

≥98%

Synonym(s):

Fluoroform, HFC-23

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHF3
CAS Number:
Molecular Weight:
70.01
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.43 (vs air)

vapor pressure

635 psi ( 21 °C)

Assay

≥98%

bp

−84 °C (lit.)

mp

−160 °C (lit.)

SMILES string

FC(F)F

InChI

1S/CHF3/c2-1(3)4/h1H

InChI key

XPDWGBQVDMORPB-UHFFFAOYSA-N

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Zanardi et al.
Journal of the American Chemical Society, 133(51), 20901-20913 (2011-12-06)
We have found the first reaction of direct cupration of fluoroform, the most attractive CF(3) source for the introduction of the trifluoromethyl group into organic molecules. Treatment of CuX (X = Cl, Br, I) with 2 equiv of MOR (M
Naleen B Jayaratna et al.
Inorganic chemistry, 52(4), 1691-1693 (2013-02-02)
Silver and copper ethylene adducts and the silver carbonyl complex of the tris(pyrazolyl)borate [HB(3,4,5-(CF(3))(3)Pz)(3)](-) (which is based on one of the most acidic pyrazoles known) have been synthesized. (13)C NMR resonance signals of metal-bound ethylene carbon atoms of [HB(3,4,5-(CF(3))(3)Pz)(3)]Ag(C(2)H(4)) and
Mark A Honey et al.
The Journal of organic chemistry, 77(3), 1396-1405 (2012-01-24)
Ethyl 2-diazo-4,4,4-trifluoro-3-oxobutanoate is a highly versatile intermediate for the synthesis of a wide range of trifluoromethyl heterocycles. With the use of rhodium(II) or copper(II) catalyzed carbene X-H insertion reactions as key steps, a diverse set of trifluoromethyl-oxazoles, -thiazoles, -imidazoles, -1,2,4-triazines
Charles S Thomoson et al.
The Journal of organic chemistry, 78(17), 8904-8908 (2013-08-13)
Fluoroform, CHF3, a non-ozone-depleting, nontoxic, and inexpensive gas can be used as a difluorocarbene source in a process for the conversion of phenols and thiophenols to their difluoromethoxy and difluorothiomethoxy derivatives. The reactions are carried out at moderate temperatures and
Shun Noritake et al.
Organic & biomolecular chemistry, 7(17), 3599-3604 (2009-08-14)
Chiral nonracemic guanidines act as Brønsted bases to generate guanidinium enolates for the enantioselective electrophilic trifluoromethylation of beta-keto esters by means of S-(trifluoromethyl)dibenzothiophenium tetrafluoroborate (Umemoto reagent) with good enantioselectivity of 60-70% range. Despite the fact that the ees are still

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service