Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

281611

Sigma-Aldrich

Tamoxifen

96%

Synonym(s):

(Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene, trans-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(C2H5)=C(C6H5)C6H4OCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
371.51
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

96%

mp

97-98 °C (lit.)

SMILES string

CC\C(c1ccccc1)=C(/c2ccccc2)c3ccc(OCCN(C)C)cc3

InChI

1S/C26H29NO/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3/h5-18H,4,19-20H2,1-3H3/b26-25-

InChI key

NKANXQFJJICGDU-QPLCGJKRSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Protein kinase C inhibitor. Induces apoptosis in human malignant glioma cell lines. Tamoxifen and its metabolite 4-hydroxytamoxifen are selective estrogen response modifiers (SERMs) that act as estrogen antagonists in mammary gland. Blocks estradiol-stimulated VEGF production in breast tumor cells.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service