Skip to Content
MilliporeSigma
Tamoxifen certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Tamoxifen

Tamoxifen certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene, trans-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethylamine, Tamoxifen
Linear Formula:
C6H5C(C2H5)=C(C6H5)C6H4OCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
371.51
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Special Grade
Technique