Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

271950

Sigma-Aldrich

Acetazolamide

99%

Synonym(s):

5-Acetamido-1,3,4-thiadiazole-2-sulfonamide, N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, N-(5-[Aminosulfonyl]-1,3,4-thiadiazol-2-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3S2
CAS Number:
Molecular Weight:
222.25
EC Number:
MDL number:

Assay

99%

SMILES string

CC(=O)Nc1nnc(s1)S(N)(=O)=O

InChI

1S/C4H6N4O3S2/c1-2(9)6-3-7-8-4(12-3)13(5,10)11/h1H3,(H2,5,10,11)(H,6,7,9)

InChI key

BZKPWHYZMXOIDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Carbonic anhydrase inhibitor; increases cerebral blood flow.

Biochem/physiol Actions

Inhibits water permeability of membranes by interacting with aquaporins

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service