Skip to Content
MilliporeSigma
Acetazolamide British Pharmacopoeia (BP) Reference Standard

Acetazolamide

Acetazolamide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
5-Acetamido-1,3,4-thiadiazole-2-sulfonamide, N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, N-(5-[Aminosulfonyl]-1,3,4-thiadiazol-2-yl)acetamide, Acetazolamide
Empirical Formula (Hill Notation):
C4H6N4O3S2
CAS Number:
Molecular Weight:
222.25

Products

Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
pK Value
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade