Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

265144

Sigma-Aldrich

2,3-Difluorobenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CHO
CAS Number:
Molecular Weight:
142.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

64-65 °C/17 mmHg (lit.)

density

1.301 g/mL at 25 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

Fc1cccc(C=O)c1F

InChI

1S/C7H4F2O/c8-6-3-1-2-5(4-10)7(6)9/h1-4H

InChI key

WDBAXYQUOZDFOJ-UHFFFAOYSA-N

General description

The emission and absorption spectra of 2,3-difluorobenzaldehyde were studied[1].

Application

2,3-Difluorobenzaldehyde was used in the synthesis of aminophosphine[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Mino et al.
The Journal of organic chemistry, 71(19), 7346-7353 (2006-09-09)
N-Aryl indoline-type aminophosphines 1a-c were obtained in good yields by a nucleophilic aromatic substitution (S(N)Ar) reaction followed by silane reduction. Aminophosphine 1d was also prepared from 2,3-difluorobenzaldehyde (4) via dimethylhydrazone. Optical resolution of C(aryl)-N(amine) bond atropisomers was achieved using (S)-(+)-di-mu-chlorobis[2-[(dimethylamino)ethyl]phenyl-C(2),N]dipalladium(II)
Spectroscopy and dynamics of the excited singlet and triplet states of 2, 3-, 2, 4-and 2, 5-difluorobenzaldehyde vapors.
Itoh, T.
Journal of Molecular Spectroscopy, 257(2), 170-176 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service