MilliporeSigma
  • Synthesis and optical resolution of aminophosphines with axially chiral C(aryl)-N(amine) bonds for use as ligands in asymmetric catalysis.
Synthesis and optical resolution of aminophosphines with axially chiral C(aryl)-N(amine) bonds for use as ligands in asymmetric catalysis.
The Journal of organic chemistry (2006-09-09)
Takashi Mino, Youichi Tanaka, Youtaro Hattori, Toshihiro Yabusaki, Hiroaki Saotome, Masami Sakamoto, Tsutomu Fujita
ABSTRACT
N-Aryl indoline-type aminophosphines 1a-c were obtained in good yields by a nucleophilic aromatic substitution (S(N)Ar) reaction followed by silane reduction. Aminophosphine 1d was also prepared from 2,3-difluorobenzaldehyde (4) via dimethylhydrazone. Optical resolution of C(aryl)-N(amine) bond atropisomers was achieved using (S)-(+)-di-mu-chlorobis[2-[(dimethylamino)ethyl]phenyl-C(2),N]dipalladium(II) ((S)-10). The determination of absolute configuration and the investigation of the rotation barrier for C(aryl)-N(amine) bond axial stability of an aminophosphine 1 are described. Finally, the ability of the chiral phosphine ligand 1 is demonstrated in a catalytic asymmetric reaction, such as a palladium-catalyzed asymmetric allylic alkylation of 1,3-diphenyl-2-propenyl acetate with dimethyl malonate (up to 95% ee).