Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

259357

Sigma-Aldrich

Bromoacetic acid

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2COOH
CAS Number:
Molecular Weight:
138.95
Beilstein:
506167
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

product line

ReagentPlus®

Assay

≥99%

bp

208 °C (lit.)

mp

47-49 °C (lit.)

SMILES string

OC(=O)CBr

InChI

1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

KDPAWGWELVVRCH-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurbir Singh et al.
Journal of colloid and interface science, 496, 278-289 (2017-02-27)
New ester-functionalized surface active ionic liquids (SAILs) based on nicotine, [C
Shi Cheng et al.
Chemosphere, 258, 127393-127393 (2020-09-20)
UV/chlorine and chlorination processes have drawn great interests of water treatment utilities for oxidation and disinfection purposes. This work proposed a restricted chlorine-dosing strategy for UV/chlorine and post-chlorination under different pH and UV irradiation conditions by comprehensively assessing the oxidation
Andrius Žilionis
European journal of mass spectrometry (Chichester, England), 26(1), 25-35 (2019-08-21)
Liquid chromatography-tandem mass spectrometry is one of the most sensitive tools for determination of trace amounts of analytes in metabolomics and proteomics. The highest sensitivity is achieved in selected reaction monitoring detection, which involves fragmentation of the molecular ion between
Avinash Bhadani et al.
Physical chemistry chemical physics : PCCP, 17(29), 19474-19483 (2015-07-07)
A new series of ester functionalized cationic gemini surfactants having different cationic headgroups (i.e. piperidinium, pyrrolidinium, morpholinium and quaternary ammonium) have been synthesized and characterized using NMR and Mass spectroscopy. These new gemini surfactants were investigated for their micellization and
Shou-Feng Wang et al.
Bioorganic & medicinal chemistry letters, 19(17), 5009-5011 (2009-08-04)
Zinc-binding groups (ZBGs) are exhaustively applied in the development of the new inhibitors against a wide variety of physiologically and pathologically important zinc proteases. Here the alpha-nitro ketone was presented as a new ZBG, which is a transition-state analog featured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service