MilliporeSigma
  • Developing a restricted chlorine-dosing strategy for UV/chlorine and post-chlorination under different pH and UV irradiation wavelength conditions.
Developing a restricted chlorine-dosing strategy for UV/chlorine and post-chlorination under different pH and UV irradiation wavelength conditions.
Chemosphere (2020-09-20)
Shi Cheng, Ji Wu, Yan-Ting Zuo, Yu-Ze Han, Wen-Xiang Ji, Yan Li, Zong-Li Huo, Ai-Min Li, Wen-Tao Li
ABSTRACT
UV/chlorine and chlorination processes have drawn great interests of water treatment utilities for oxidation and disinfection purposes. This work proposed a restricted chlorine-dosing strategy for UV/chlorine and post-chlorination under different pH and UV irradiation conditions by comprehensively assessing the oxidation of natural organic matter (NOM), formation of 9 haloacetic acids (HAA9) and bromate, and alteration of toxicity. During UV/chlorine with restricted chlorine doses, the oxidation of NOM chromophores (i.e., ΔUVA254) showed an apparent dependence on cumulative exposures of free available chlorine (CTFAC); Meanwhile, HAA9 formation was determined by CTFAC values and could be linearly correlated with ΔUVA254 irrespective of pH and UV irradiation wavelength. Irradiated by 254 nm LP-Hg lamp, the faster chlorine photolysis produced relatively higher steady-state concentrations of Cl• and HO• species but resulted in lower CTFAC. Reducing CTFAC values by operation parameters (pH, UV wavelength and irradiation fluence) could mitigate HAA9 formation during UV/chlorine at a specific chlorine dose. Additionally, high bromide concentration and acidic pH promoted more bromo-HAAs formation, and the presence of NOM significantly suppressed bromate formation. Analogous to ozonation, the UV/chlorine pre-oxidation could reduce the HAA9 formation potentials during post-chlorination at mildly alkaline pH. The photobacterium bioassay further demonstrated that although the UV/chlorine treatment might have increased the acute toxicity, the post-chlorination treatment could polish the acute toxicity to the level of chlorination alone. These results suggest that with the restricted chlorine-dosing strategy, the trade-off between oxidation/disinfection efficiency and DBPs formation can be controlled by monitoring CTFAC and ΔUVA254 values during UV/chlorine treatment.
Sigma-Aldrich
Tribromoacetic acid, 99%
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O