Skip to Content
MilliporeSigma
All Photos(1)

Documents

239372

Sigma-Aldrich

3-Amino-2-cyclohexen-1-one

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H7O
CAS Number:
Molecular Weight:
111.14
Beilstein:
471354
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

129-133 °C (lit.)

SMILES string

NC1=CC(=O)CCC1

InChI

1S/C6H9NO/c7-5-2-1-3-6(8)4-5/h4H,1-3,7H2

InChI key

ZZMRPOAHZITKBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Amino-2-cyclohexen-1-one was used in the synthesis of a series of 2-amino-5-oxo-4-phenyl-5,6,7,8-tetrahydroquinoline-3-carbonitriles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrice L Jackson et al.
Bioorganic & medicinal chemistry, 17(1), 133-140 (2008-12-09)
3D-QSAR studies comparative molecular field analysis (CoMFA) and comparative molecular similarity indices analysis (CoMSIA) were carried out on 26 structurally diverse subcutaneous pentylenetetrazol (scPTZ) active enaminone analogues, previously synthesized in our laboratory. CoMFA and CoMSIA were employed to generate models
Atul R Gholap et al.
Bioorganic & medicinal chemistry, 15(21), 6705-6715 (2007-09-04)
A series of 2-amino-5-oxo-4-phenyl-5,6,7,8-tetrahydroquinoline-3-carbonitrile and various analogues have been synthesized in excellent isolated yields starting from various arylidenemalononitrile and 3-amino-2-cyclohexen-1-one in 1-propanol as solvent at reflux temperature in the absence of any added catalyst. All the synthesized compounds were evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service