MilliporeSigma
  • Synthesis and evaluation of antifungal properties of a series of the novel 2-amino-5-oxo-4-phenyl-5,6,7,8-tetrahydroquinoline-3-carbonitrile and its analogues.
Synthesis and evaluation of antifungal properties of a series of the novel 2-amino-5-oxo-4-phenyl-5,6,7,8-tetrahydroquinoline-3-carbonitrile and its analogues.
Bioorganic & medicinal chemistry (2007-09-04)
Atul R Gholap, Kiran S Toti, Fazal Shirazi, Ratna Kumari, Manoj Kumar Bhat, Mukund V Deshpande, Kumar V Srinivasan
ABSTRACT
A series of 2-amino-5-oxo-4-phenyl-5,6,7,8-tetrahydroquinoline-3-carbonitrile and various analogues have been synthesized in excellent isolated yields starting from various arylidenemalononitrile and 3-amino-2-cyclohexen-1-one in 1-propanol as solvent at reflux temperature in the absence of any added catalyst. All the synthesized compounds were evaluated for their antifungal activity. The relationship between functional group variation and biological activity of the evaluated compounds is discussed in the article.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture