Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

222623

Sigma-Aldrich

2,5-Dimethyl-3-hexyne-2,5-diol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(CH3)2C≡CC(CH3)2OH
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

bp

121-123 °C/7 mmHg (lit.)

mp

90-94 °C (lit.)

SMILES string

CC(C)(O)C#CC(C)(C)O

InChI

1S/C8H14O2/c1-7(2,9)5-6-8(3,4)10/h9-10H,1-4H3

InChI key

IHJUECRFYCQBMW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service