Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

215201

Sigma-Aldrich

2-Hydroxyethylhydrazine

technical grade, 90%

Synonym(s):

2-Hydrazinoethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2NHNH2
CAS Number:
Molecular Weight:
76.10
Beilstein:
1731669
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

Assay

90%

refractive index

n20/D 1.493 (lit.)

bp

155-160 °C/32 mmHg (lit.)

mp

−70 °C (lit.)

density

1.123 g/mL at 25 °C (lit.)

SMILES string

NNCCO

InChI

1S/C2H8N2O/c3-4-1-2-5/h4-5H,1-3H2

InChI key

GBHCABUWWQUMAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory M Sandala et al.
Journal of the American Chemical Society, 127(24), 8856-8864 (2005-06-16)
Ab initio molecular orbital calculations have been used to study the mechanism of suicide inactivation of ethanolamine ammonia-lyase induced by three different substrate analogues. Analysis of the normal catalytic mechanism with 2-aminoethanol (ethanolamine) as substrate predicts that both the hydrogen-abstraction
R D Slocum et al.
Planta, 183(3), 443-450 (1991-02-01)
An electron-microscopic cytochemical method was used to localize diamine oxidase (DAO) in pea and polyamine oxidase (PAO) in maize (Zea mays L.). The method, based on the precipitation of amine-oxidase-generated H2O2 by CeCl3, was shown to be specific for DAO
A M Timperio et al.
Xenobiotica; the fate of foreign compounds in biological systems, 33(2), 153-167 (2003-03-08)
1 Furazolidone, a drug widely used in human and veterinary medicine, exhibits inhibition of monoamine oxidase activity, as observed in the tissues of a number of different animal species, including man. The aim of the current study was to determine
Guoxing Su et al.
Journal of plant physiology, 162(12), 1297-1303 (2006-01-21)
In order to analyze the relationship between polyamine oxidative degradation induced by light and the Lignin synthesis in cell walls, the activities of diamine oxidases and peroxidase, the contents of H2O2 and lignin, and the growth of hypocotyls in soybean
C Andriamampandry et al.
Journal of neurochemistry, 56(6), 1845-1850 (1991-06-01)
The sequential methylation of ethanolamine and its phosphorylated derivatives has been studied with chick neurons in culture in the presence of several pharmacological agents. Incubation with [3H]ethanolamine in the presence of monomethylethanolamine and dimethylethanolamine indicated that in these neurons the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service