MilliporeSigma
  • Electron-microscopic cytochemical localization of diamine and polyamine oxidases in pea and maize tissues.
Electron-microscopic cytochemical localization of diamine and polyamine oxidases in pea and maize tissues.
Planta (1991-02-01)
R D Slocum, M J Furey
ABSTRACT
An electron-microscopic cytochemical method was used to localize diamine oxidase (DAO) in pea and polyamine oxidase (PAO) in maize (Zea mays L.). The method, based on the precipitation of amine-oxidase-generated H2O2 by CeCl3, was shown to be specific for DAO and PAO and permitted their localization in plant tissues with a high degree of resolution. Both enzymes are localized exclusively in the cell wall. Both DAO- and PAO-activity staining is most intense in the middle lamellar region of the wall and in cells exhibiting highly lignified walls. The oxidases could provide H2O2 for peroxidase-mediated cross-linking reactions in the cell wall and may, in this capacity, play a role in the regulation of plant growth.
Sigma-Aldrich
2-Hydroxyethylhydrazine, ≥95% (GC)