Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

209805

Sigma-Aldrich

3-(1-Cyanoethyl)benzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(CN)C6H4CO2H
CAS Number:
Molecular Weight:
175.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

145-148 °C (lit.)

functional group

carboxylic acid
nitrile

SMILES string

CC(C#N)c1cccc(c1)C(O)=O

InChI

1S/C10H9NO2/c1-7(6-11)8-3-2-4-9(5-8)10(12)13/h2-5,7H,1H3,(H,12,13)

InChI key

IRYIYPWRXROPSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(1-Cyanoethyl)benzoic acid was used in screening of cobalt containing nitrile hydratases for their enantioselectivity towards wide range of chiral nitriles.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S van Pelt et al.
Organic & biomolecular chemistry, 9(8), 3011-3019 (2011-03-08)
In this study a diverse range of purified cobalt containing nitrile hydratases (NHases, EC 4.2.1.84) from Rhodopseudomonas palustris HaA2 (HaA2), Rhodopseudomonas palustris CGA009 (009), Sinorhizobium meliloti 1021 (1021), and Nitriliruptor alkaliphilus (iso2), were screened for the first time for their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service