Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

208744

Sigma-Aldrich

(2-Fluorophenyl)acetone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2COCH3
CAS Number:
Molecular Weight:
152.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.4989 (lit.)

bp

47 °C/0.05 mmHg (lit.)

density

1.077 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Cc1ccccc1F

InChI

1S/C9H9FO/c1-7(11)6-8-4-2-3-5-9(8)10/h2-5H,6H2,1H3

InChI key

BANVZEUCJHUPOI-UHFFFAOYSA-N

General description

(2-Fluorophenyl)acetone on reaction with chlorosulfonyl isocyanate yields 5-(2-fluorophenyl)-6-methyl-1,3-oxazine-2,4-(3H)-dione.

Application

(2-Fluorophenyl)acetone was used in the synthesis of 6-methyluracils.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer A Weiß et al.
Science & justice : journal of the Forensic Science Society, 57(1), 6-12 (2017-01-09)
Amphetamine and its derivatives gained high popularity on the illegal drug market. In the last few years, a lot of new psychoactive compounds structurally related to amphetamine, such as 4-fluoroamphetamine and 4-fluoromethamphetamine swamped the drug market. They were designed to
Martin W Rowbottom et al.
Bioorganic & medicinal chemistry letters, 14(9), 2269-2274 (2004-04-15)
The synthesis of a series of (R)-1-alkyl-3-[2-(2-amino)phenethyl]-5-(2-fluorophenyl)-6-methyluracils is discussed. SAR around N-1 of the uracil was explored, which led to the discovery that an electron-deficient 2,6-disubstituted benzyl group is required for optimal receptor binding. The best compound from the series

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service