MilliporeSigma
  • Synthesis and structure-activity relationships of (R)-1-alkyl-3-[2-(2-amino)phenethyl]-5-(2-fluorophenyl)-6-methyluracils as human GnRH receptor antagonists.
Synthesis and structure-activity relationships of (R)-1-alkyl-3-[2-(2-amino)phenethyl]-5-(2-fluorophenyl)-6-methyluracils as human GnRH receptor antagonists.
Bioorganic & medicinal chemistry letters (2004-04-15)
Martin W Rowbottom, Fabio C Tucci, Yun-Fei Zhu, Zhiqiang Guo, Timothy D Gross, Greg J Reinhart, Qui Xie, R Scott Struthers, John Saunders, Chen Chen
ABSTRACT
The synthesis of a series of (R)-1-alkyl-3-[2-(2-amino)phenethyl]-5-(2-fluorophenyl)-6-methyluracils is discussed. SAR around N-1 of the uracil was explored, which led to the discovery that an electron-deficient 2,6-disubstituted benzyl group is required for optimal receptor binding. The best compound from the series had binding affinity of 0.7 nM (K(i) for the human GnRH receptor, which was 8-fold better than the 2,6-difluorobenzyl analog.
Sigma-Aldrich
6-Methyluracil, 97%