Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

203750

Sigma-Aldrich

Manganese(IV) oxide

99.99% trace metals basis

Synonym(s):

Manganese dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38190113
PubChem Substance ID:

Assay

99.99% trace metals basis

form

solid

mp

535 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

O=[Mn]=O

InChI

1S/Mn.2O

InChI key

NUJOXMJBOLGQSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemical insertion of lithium into this product formed a spinel of high capacity for added lithium insertion and ion exchange.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammundsen, B. et al.
Chemistry of Materials, 10, 1680-1680 (1998)
Xing Liu et al.
The Analyst, 137(19), 4552-4558 (2012-08-18)
Inorganic nanomaterials that mimic enzymes are fascinating as they potentially have improved properties relative to native enzymes, such as greater resistance to extremes of pH and temperature and lower sensitivity to proteases. Although many artificial enzymes have been investigated, searching
Highly stretchable alkaline batteries based on an embedded conductive fabric.
Abhinav M Gaikwad et al.
Advanced materials (Deerfield Beach, Fla.), 24(37), 5071-5076 (2012-07-05)
Xin Zhao et al.
ACS nano, 6(6), 5404-5412 (2012-05-05)
Manganese dioxide (MnO(2)) particles 2-3 nm in size were deposited onto a porous "activated microwave expanded graphite oxide" (aMEGO) carbon scaffold via a self-controlled redox process. Symmetric electrochemical capacitors were fabricated that yielded a specific capacitance of 256 F/g (volumetric:
Y Wang et al.
Journal of colloid and interface science, 380(1), 8-15 (2012-06-02)
Bio-inspired chemical approach has been developed for the surface modification and electrophoretic deposition of manganese dioxide and zirconia nanoparticles, prepared by chemical precipitation methods. Caffeic acid, trans-cinnamic acid, p-coumaric acid, and 2,4-dihydroxycinnamic acid were investigated for the surface modification of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service