MilliporeSigma
  • Incorporation of manganese dioxide within ultraporous activated graphene for high-performance electrochemical capacitors.
Incorporation of manganese dioxide within ultraporous activated graphene for high-performance electrochemical capacitors.
ACS nano (2012-05-05)
Xin Zhao, Lili Zhang, Shanthi Murali, Meryl D Stoller, Qinghua Zhang, Yanwu Zhu, Rodney S Ruoff
ABSTRACT
Manganese dioxide (MnO(2)) particles 2-3 nm in size were deposited onto a porous "activated microwave expanded graphite oxide" (aMEGO) carbon scaffold via a self-controlled redox process. Symmetric electrochemical capacitors were fabricated that yielded a specific capacitance of 256 F/g (volumetric: 640 F/cm(3)) and a capacitance retention of 87.7% after 1000 cycles in 1 M H(2)SO(4); when normalized to MnO(2), the specific capacitance was 850 F/g. Asymmetric electrochemical capacitors were also fabricated with aMEGO/MnO(2) as the positive electrode and aMEGO as the negative electrode and had a power density of 32.3 kW/kg (for an energy density of 20.8 Wh/kg), an energy density of 24.3 Wh/kg (for a power density of 24.5 kW/kg), and a capacitance retention of 80.5% over 5000 cycles.
Sigma-Aldrich
Manganese(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Manganese(IV) oxide, activated, ~85%, <10 μm
Sigma-Aldrich
Manganese(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(IV) oxide, 10 μm, reagent grade, ≥90%