Skip to Content
MilliporeSigma
All Photos(1)

Documents

197971

Sigma-Aldrich

Ethyl homovanillate

97%

Synonym(s):

Ethyl 4-hydroxy-3-methoxyphenylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH2CO2C2H5
CAS Number:
Molecular Weight:
210.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

180-185 °C/14 mmHg (lit.)

mp

44-47 °C (lit.)

SMILES string

CCOC(=O)Cc1ccc(O)c(OC)c1

InChI

1S/C11H14O4/c1-3-15-11(13)7-8-4-5-9(12)10(6-8)14-2/h4-6,12H,3,7H2,1-2H3

InChI key

AWPMWZHWVKXADV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl homovanillate is a strong inhibitor of monoamine oxidase A (MAOA).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoxin Tao et al.
Bioorganic & medicinal chemistry, 13(15), 4777-4788 (2005-06-07)
Eugenol (1) is an active principle of Rhizoma acori graminei, a medicinal herb used in Asia for the treatment of symptoms reminiscent of Alzheimer's disease (AD). It has been shown to protect neuronal cells from the cytotoxic effect of amyloid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service