Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20611

Sigma-Aldrich

Guanidinium carbonate

for synthesis

Synonym(s):

Guanidinium carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$65.80
1 KG
$89.50
25 KG
$1,180.00
50 KG
$2,030.00

$65.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$65.80
1 KG
$89.50
25 KG
$1,180.00
50 KG
$2,030.00

About This Item

Linear Formula:
(CH5N3)2 · H2CO3
CAS Number:
Molecular Weight:
180.17
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-813-7
NACRES:
NA.22

$65.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

autoignition temp.

530 °C

potency

1045 mg/kg LD50, oral (Rat)

pH

11.7 (20 °C, 110 g/L in H2O)

mp

231 °C

solubility

450 g/L

density

1.29 g/cm3 at 22 °C

bulk density

500‑700 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3]\C(=N\[H])\N.[N+H3]\C(=N\[H])\N.[O-]C(=O)[O-]

InChI

1S/2CH5N3.CH2O3/c3*2-1(3)4/h2*(H5,2,3,4);(H2,2,3,4)

InChI key

STIAPHVBRDNOAJ-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service