Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

192201

Sigma-Aldrich

2,4,6-Trimethylbenzenesulfonohydrazide

97%

Synonym(s):

2-Mesitylenesulfonohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2SO2NHNH2
CAS Number:
Molecular Weight:
214.28
Beilstein:
2118380
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

112-114 °C (dec.) (lit.)

functional group

hydrazine

SMILES string

Cc1cc(C)c(c(C)c1)S(=O)(=O)NN

InChI

1S/C9H14N2O2S/c1-6-4-7(2)9(8(3)5-6)14(12,13)11-10/h4-5,11H,10H2,1-3H3

InChI key

JQUBKTQDNVZHIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4,6-Trimethylbenzenesulfonohydrazide has been used in the synthesis of:
  • (E)-N′-(1-((1S*,4aR*,8aR*)-1-hydroxydecahydronaphthalen-4a-yl)ethylidene)-2,4,6-trimethylbenzenesulfonohydrazide
  • complanadine alkaloids

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concise Synthesis of 1, 4a-Bifunctionalised Decalin Building Blocks by CH Activation of Decalin.
Uosis-Martin M, et al.
Synlett, 2011(15), 2211-2213 (2011)
Mario Uosis-Martin et al.
The Journal of organic chemistry, 78(12), 6253-6263 (2013-05-30)
A synthetic approach to complanadine alkaloids is described which employs a Kondrat'eva reaction to construct the pyridine rings. The viability of this approach is demonstrated by its application to a model substrate accessed from unfunctionalized decalin. The key transformation affords

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service