- A model system for the synthesis of complanadine alkaloids by "diverted Kondrat'eva" oxazole-olefin cycloaddition.
A model system for the synthesis of complanadine alkaloids by "diverted Kondrat'eva" oxazole-olefin cycloaddition.
The Journal of organic chemistry (2013-05-30)
Mario Uosis-Martin, G Dan Pantoş, Mary F Mahon, Simon E Lewis
PMID23713565
ABSTRACT
A synthetic approach to complanadine alkaloids is described which employs a Kondrat'eva reaction to construct the pyridine rings. The viability of this approach is demonstrated by its application to a model substrate accessed from unfunctionalized decalin. The key transformation affords the desired tetracyclic architecture with unprecedented incorporation of substituents on the pyridine ring, implicating the oxazole α-hydroxy group as an active participant in the cycloadduct fragmentation process.