Skip to Content
MilliporeSigma
All Photos(1)

Documents

166731

Sigma-Aldrich

Methyl 2,5-dichlorobenzoate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CO2CH3
CAS Number:
Molecular Weight:
205.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

37-40 °C (lit.)

SMILES string

COC(=O)c1cc(Cl)ccc1Cl

InChI

1S/C8H6Cl2O2/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4H,1H3

InChI key

SPJQBGGHUDNAIC-UHFFFAOYSA-N

Application

Methyl 2,5-dichlorobenzoate was used as starting reagent in the synthesis of poly(p-phenylene). It was used in isolation of microorganism degrading methyl benzoate, Burkholderia cepacia and its ability to degrade different methyl aromatic esters.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of poly (p-phenylene) via a new precursor route.
Chaturvedi V, et al.
Macromolecules, 26(10), 2607-2611 (1993)
Microbial hydrolysis of methyl aromatic esters by Burkholderia cepacia isolated from soil.
Philippe G, et al.
FEMS Microbiology Ecology, 37(3), 251-258 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service