Skip to Content
MilliporeSigma
All Photos(1)

Documents

160466

Sigma-Aldrich

1,2,5,6-Diepoxycyclooctane

96%

Synonym(s):

2,7-Dioxatwistane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O2
CAS Number:
Molecular Weight:
140.18
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

powder

refractive index

n20/D 1.496 (lit.)

bp

70 °C/0.04 mmHg (lit.)

density

1.138 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CC2OC2CCC3OC13

InChI

1S/C8H12O2/c1-2-6-8(10-6)4-3-7-5(1)9-7/h5-8H,1-4H2

InChI key

USGYMDAUQBQWFU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service