Skip to Content
MilliporeSigma
All Photos(1)

Documents

157465

Sigma-Aldrich

4′-Methoxychalcone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOC6H4OCH3
CAS Number:
Molecular Weight:
238.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

101-103 °C (lit.)

SMILES string

COc1ccc(cc1)C(=O)\C=C\c2ccccc2

Gene Information

human ... IL1B(3553)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Wattenberg
Journal of cellular biochemistry. Supplement, 22, 162-168 (1995-01-01)
The objective of the studies reported here has been to find novel chemopreventive agents effective against carcinogenesis of the lung. In particular, identification of suppressing agents, i.e., compounds preventing the evolution of the neoplastic process, has been sought. For this
Alessandra M T Souza et al.
Current microbiology, 59(4), 374-379 (2009-07-25)
Current drugs for treating leishmaniasis are still associated with significant toxicity and failure rates. Thus, new effective and less toxic antileishmanial agents are still in need. Herein, we tested a series of sulfonamide 4-methoxychalcone derivatives against L. amazonensis promastigote and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service