Skip to Content
MilliporeSigma
All Photos(1)

Documents

15066

Sigma-Aldrich

Boc-Asp-OBzl

≥98.0% (TLC)

Synonym(s):

Boc-L-aspartic acid 1-benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
323.34
Beilstein:
2481680
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CC(O)=O)C(=O)OCc1ccccc1

InChI

1S/C16H21NO6/c1-16(2,3)23-15(21)17-12(9-13(18)19)14(20)22-10-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,17,21)(H,18,19)/t12-/m0/s1

InChI key

LDRWTKQWSXGSTM-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service