Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15106

Sigma-Aldrich

Boc-D-Glu-OBzl

≥98.0% (TLC)

Synonym(s):

Boc-D-glutamic acid 1-benzyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$483.00

$483.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$483.00

About This Item

Empirical Formula (Hill Notation):
C17H23NO6
CAS Number:
Molecular Weight:
337.37
Beilstein:
4154458
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$483.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@H](CCC(O)=O)C(=O)OCc1ccccc1

InChI

1S/C17H23NO6/c1-17(2,3)24-16(22)18-13(9-10-14(19)20)15(21)23-11-12-7-5-4-6-8-12/h4-8,13H,9-11H2,1-3H3,(H,18,22)(H,19,20)/t13-/m1/s1

InChI key

CVZUKWBYQQYBTF-CYBMUJFWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service