Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

132675

Sigma-Aldrich

3-Bromostyrene

97%, contains 0.1% 3,5-di-tert-butylcatechol as inhibitor

Synonym(s):

1-Bromo-3-ethenylbenzene, 1-Bromo-3-vinylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$139.00

$139.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$139.00

About This Item

Linear Formula:
H2C=CHC6H4Br
CAS Number:
Molecular Weight:
183.05
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$139.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

contains

0.1% 3,5-di-tert-butylcatechol as inhibitor

refractive index

n20/D 1.591 (lit.)

bp

74-75 °C/3 mmHg (lit.)

density

1.406 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Brc1cccc(C=C)c1

InChI

1S/C8H7Br/c1-2-7-4-3-5-8(9)6-7/h2-6H,1H2

InChI key

KQJQPCJDKBKSLV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service