Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

369608

Sigma-Aldrich

4-(Trifluoromethyl)styrene

98%, contains 0.1% 4-tert-butylcatechol as inhibitor

Synonym(s):

4-Vinylbenzotrifluoride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$213.00

$213.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$213.00

About This Item

Linear Formula:
CF3C6H4CH=CH2
CAS Number:
Molecular Weight:
172.15
Beilstein:
1863548
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$213.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

contains

0.1% 4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.466 (lit.)

bp

65-66 °C/40 mmHg (lit.)

density

1.165 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)c1ccc(C=C)cc1

InChI

1S/C9H7F3/c1-2-7-3-5-8(6-4-7)9(10,11)12/h2-6H,1H2

InChI key

CEWDRCQPGANDRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service