Skip to Content
MilliporeSigma
All Photos(1)

Documents

127264

Sigma-Aldrich

2-Norbornaneacetic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O2
CAS Number:
Molecular Weight:
154.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.4827 (lit.)

bp

107-108 °C/3 mmHg (lit.)

density

1.065 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CC1C[C@@H]2CC[C@H]1C2

InChI

1S/C9H14O2/c10-9(11)5-8-4-6-1-2-7(8)3-6/h6-8H,1-5H2,(H,10,11)/t6-,7+,8?/m1/s1

InChI key

FYHBMPWRHCWNBC-KVARREAHSA-N

Application

2-Norbornaneacetic acid was used to study the synthesis and antibacterial activity (structure) of triazolyl oxazolidinones.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oludotun A Phillips et al.
European journal of medicinal chemistry, 43(5), 1095-1104 (2008-02-05)
A series of new piperazinyl 5-triazolylmethyl oxazolidinones containing long chain acyl group at the piperazine N-4-position were synthesized and evaluated against a panel of standard and clinical isolates of Gram-positive and Gram-negative bacteria. Derivatives having long chain acyl groups with
Michael J Wilde et al.
Journal of chromatography. A, 1378, 74-87 (2015-01-03)
Although bicyclic acids have been reported to be the major naphthenic acids in oil sands process-affected water (OSPW) and a well-accepted screening assay indicated that some bicyclics were the most acutely toxic acids tested, none have yet been identified. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service