Skip to Content
MilliporeSigma
All Photos(1)

Documents

772828

Sigma-Aldrich

5-Norbornene-2-endo-acetic acid

Synonym(s):

Bicyclo[2.2.1]hept-5-en-2-ylacetic acid, Norbornene acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O2
CAS Number:
Molecular Weight:
152.19
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: linker

refractive index

n20/D 1.495

density

1.124 g/mL at 25 °C

functional group

carboxylic acid

SMILES string

OC(CC1C[C@@H]2C=C[C@H]1C2)=O

InChI

1S/C9H12O2/c10-9(11)5-8-4-6-1-2-7(8)3-6/h1-2,6-8H,3-5H2,(H,10,11)/t6-,7+,8?/m1/s1

InChI key

HRVGJQMCNYJEHM-KVARREAHSA-N

Application

Carboxylic acid functionalized norbornene. Carboxylic acid can be used to modify amine containing compounds such as lysine residues on proteins/peptides via standard peptide coupling methods. Norbornene has been shown to react with 1,2,4,5-tetrazines in an inverse electron demand Diels-Alder cycloaddition reaction, which is a bioorthogonal click chemistry reaction demonstrated to be useful in biological imaging applications.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee-Sun Han et al.
Journal of the American Chemical Society, 132(23), 7838-7839 (2010-05-21)
We present a bioorthogonal and modular conjugation method for efficient coupling of organic dyes and biomolecules to quantum dots (QDs) using a norbornene-tetrazine cycloaddition. The use of noncoordinating functional groups combined with the rapid rate of the cycloaddition leads to
Neal K Devaraj et al.
Bioconjugate chemistry, 19(12), 2297-2299 (2008-12-05)
Bioorthogonal tetrazine cycloadditions have been applied to live cell labeling. Tetrazines react irreversibly with the strained dienophile norbornene forming dihydropyrazine products and dinitrogen. The reaction is high yielding, selective, and fast in aqueous media. Her2/neu receptors on live human breast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service