Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

123595

Sigma-Aldrich

2,3-Benzofluorene

98%

Synonym(s):

11H-Benzo[b]fluorene, Isonaphthofluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12
CAS Number:
Molecular Weight:
216.28
Beilstein:
2046365
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

211-213 °C (lit.)

solubility

methanol: soluble

SMILES string

C1c2ccccc2-c3cc4ccccc4cc13

InChI

1S/C17H12/c1-2-6-13-11-17-15(9-12(13)5-1)10-14-7-3-4-8-16(14)17/h1-9,11H,10H2

InChI key

HAPOJKSPCGLOOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Benzofluorene was identified in a coal liquid (solvent-refined coal) product by synchronous fluorescence and the room-temperature phosphorescence methods[1]. It inhibits estradiol-dependent reporter activity in yeast[2].

Application

2,3-Benzofluorene was used in three way analysis of fluorescence excitation-emission matrices of mixtures of polycyclic aromatic hydrocarbons[3]. It was used as standard to detect the ambient particle-bound polycyclic aromatic hydrocarbons by a real-time aerosol mass spectrometer[4].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Staicu et al.
The Journal of chemical physics, 129(7), 074302-074302 (2008-12-03)
The S(1)((1)A('))<--S(0)((1)A(')) absorption spectrum of jet-cooled 2,3-benzofluorene (Bzf) has been measured by cavity ring-down spectroscopy. The potential energy surfaces of the S(n=0,1,2) states of Bzf have been investigated with calculations based on the time-dependent density functional theory (TD-DFT). At the
D Q Tran et al.
Biochemical and biophysical research communications, 229(1), 101-108 (1996-12-04)
Polynuclear aromatic hydrocarbons (PAH) represent a large class of chemicals present in environment. We have used yeast strain ER(wt) expressing human estrogen receptor (hER) and an estrogen-sensitive reporter to characterize the estrogenic or anti-estrogenic activities of 21 PAHs. The PAHs
Direct determination of selected polynuclear aromatic hydrocarbons in a coal liquefaction product by synchronous luminescence techniques.
Vo-Dinh T and Martinez PR.
Analytica Chimica Acta, 125, 13-19 (1981)
Shugao Zhu et al.
Organic & biomolecular chemistry, 10(18), 3696-3704 (2012-04-13)
We report in this paper an interesting tandem reaction involving sequential palladium(0)-catalyzed decarboxylation of diynylic carbonates, intramolecular nucleophilic cyclization and Schmittel reaction, which provides a facile method for the synthesis of a variety of polycyclic benzo[b]fluorene derivatives from easily accessible
Detection of particle-phase polycyclic aromatic hydrocarbons in Mexico City using an aerosol mass spectrometer.
Dzepina K, et al.
International Journal of Mass Spectrometry, 263(2), 152-170 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service