Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

261424

Sigma-Aldrich

Cyclopenta[d,e,f]phenanthrene

97%

Synonym(s):

4,5-Methylenephenanthrene

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$360.00

$360.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C15H10
CAS Number:
Molecular Weight:
190.24
Beilstein:
2043908
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$360.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

bp

353 °C (lit.)

mp

113-115 °C (lit.)

SMILES string

C1c2cccc3ccc4cccc1c4c23

InChI

1S/C15H10/c1-3-10-7-8-11-4-2-6-13-9-12(5-1)14(10)15(11)13/h1-8H,9H2

InChI key

RKZDZWJDQTZDLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclopenta[d,e,f]phenanthrene (4,5-methylenephenanthrene) has been used as an indicator in the synthesis of homopolymers and block copolymers of styrene and dienes with variable 1,2-polydiene content[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simplified procedure of anionic polymerization of styrene and dienes using 4, 5-methylenephenanthrene as an indicator.
Sanger J, et al.
Polymer Bull., 36(1), 19-26 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service