Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

122912

Sigma-Aldrich

2-Amino-6,8-dihydroxypurine

Synonym(s):

2-Amino-6,8-purinediol, 8-Hydroxyguanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5O2
CAS Number:
Molecular Weight:
167.13
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

impurities

<10% acetic acid

SMILES string

Nc1nc(O)c2[nH]c(O)nc2n1

InChI

1S/C5H5N5O2/c6-4-8-2-1(3(11)10-4)7-5(12)9-2/h(H5,6,7,8,9,10,11,12)

InChI key

CLGFIVUFZRGQRP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Guo et al.
Nucleic acids research, 41(16), 7700-7712 (2013-06-19)
Oxidized bases in DNA have been implicated in cancer, aging and neurodegenerative disease. We have developed an approach combining single-cell gel electrophoresis (comet) with fluorescence in situ hybridization (FISH) that enables the comparative quantification of low, physiologically relevant levels of
Ana-Maria Chiorcea-Paquim et al.
Mikrochimica acta, 188(2), 58-58 (2021-01-29)
Oxidative DNA damage plays an important role in the pathogenesis of various diseases. Among oxidative DNA lesions, 8-oxoguanine (8-oxoG) and its corresponding nucleotide 8-oxo-2'-deoxyguanosine (8-oxodG), the guanine and deoxyguanosine oxidation products, have gained much attention, being considered biomarkers for oxidative
Ilinca Nicolae et al.
Experimental and therapeutic medicine, 17(2), 1012-1019 (2019-01-27)
Recent data suggest that severe psoriasis is an independent risk factor for chronic renal disease. In the present study, we investigated the role of related-purine derivatives as predictors of renal dysfunctions in patients with psoriasis. A prospective study was conducted
Maura Costello et al.
Nucleic acids research, 41(6), e67-e67 (2013-01-11)
As researchers begin probing deep coverage sequencing data for increasingly rare mutations and subclonal events, the fidelity of next generation sequencing (NGS) laboratory methods will become increasingly critical. Although error rates for sequencing and polymerase chain reaction (PCR) are well
Sara Timpano et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(4), 5716-5728 (2019-01-17)
Multicellular organisms balance oxygen delivery and toxicity by having oxygen pass through several barriers before cellular delivery. In human cell culture, these physiologic barriers are removed, exposing cells to higher oxygen levels. Human cells cultured in ambient air may appear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service