MilliporeSigma
  • Nanostructured material-based electrochemical sensing of oxidative DNA damage biomarkers 8-oxoguanine and 8-oxodeoxyguanosine: a comprehensive review.
Nanostructured material-based electrochemical sensing of oxidative DNA damage biomarkers 8-oxoguanine and 8-oxodeoxyguanosine: a comprehensive review.
Mikrochimica acta (2021-01-29)
Ana-Maria Chiorcea-Paquim, Ana Maria Oliveira-Brett
ABSTRACT
Oxidative DNA damage plays an important role in the pathogenesis of various diseases. Among oxidative DNA lesions, 8-oxoguanine (8-oxoG) and its corresponding nucleotide 8-oxo-2'-deoxyguanosine (8-oxodG), the guanine and deoxyguanosine oxidation products, have gained much attention, being considered biomarkers for oxidative DNA damage. Both 8-oxoG and 8-oxodG are used to predict overall body oxidative stress levels, to estimate the risk, to detect, and to make prognosis related to treatment of cancer, degenerative, and other age-related diseases. The need for rapid, easy, and low-cost detection and quantification of 8-oxoG and 8-oxodG biomarkers of oxidative DNA damage in complex samples, urine, blood, and tissue, caused an increasing interest on electrochemical sensors based on modified electrodes, due to their high sensitivity and selectivity, low-cost, and easy miniaturization and automation. This review aims to provide a comprehensive and exhaustive overview of the fundamental principles concerning the electrochemical determination of the biomarkers 8-oxoG and 8-oxodG using nanostructured materials (NsM), such as carbon nanotubes, carbon nanofibers, graphene-related materials, gold nanomaterials, metal nanoparticles, polymers, nanocomposites, dendrimers, antibodies and aptamers, and modified electrochemical sensors.