Skip to Content
MilliporeSigma
All Photos(1)

Documents

105635

Sigma-Aldrich

Methyl 1-methylcyclopropyl ketone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C3H4COCH3
CAS Number:
Molecular Weight:
98.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.434 (lit.)

bp

125-128 °C (lit.)

density

0.895 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C1(C)CC1

InChI

1S/C6H10O/c1-5(7)6(2)3-4-6/h3-4H2,1-2H3

InChI key

OQBCJXUAQQMTRW-UHFFFAOYSA-N

General description

Methyl 1-methylcyclopropyl ketone undergoes samarium diiodide induced coupling to form 6-Hydroxy-3-methyl-6-(1-methylcyclopropyl)heptan-2-one.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samarium Diiodide Induced Reactions of Cyclopropyl Ketones: Reductive Ring Cleavage and Dimerization Leading to 1, 8-Diketones-Scope, Limitations, Mechanisms.
Aulenta F, et al.
European Journal of Organic Chemistry, 2006(7), 1733-1739 (2006)
Valeriy V Pak et al.
Cell metabolism, 31(3), 642-653 (2020-03-05)
Hydrogen peroxide (H2O2) is a key redox intermediate generated within cells. Existing probes for H2O2 have not solved the problem of detection of the ultra-low concentrations of the oxidant: these reporters are not sensitive enough, or pH-dependent, or insufficiently bright

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service