MilliporeSigma
  • Ultrasensitive Genetically Encoded Indicator for Hydrogen Peroxide Identifies Roles for the Oxidant in Cell Migration and Mitochondrial Function.
Ultrasensitive Genetically Encoded Indicator for Hydrogen Peroxide Identifies Roles for the Oxidant in Cell Migration and Mitochondrial Function.
Cell metabolism (2020-03-05)
Valeriy V Pak, Daria Ezeriņa, Olga G Lyublinskaya, Brandán Pedre, Pyotr A Tyurin-Kuzmin, Natalie M Mishina, Marion Thauvin, David Young, Khadija Wahni, Santiago Agustín Martínez Gache, Alexandra D Demidovich, Yulia G Ermakova, Yulia D Maslova, Arina G Shokhina, Emrah Eroglu, Dmitry S Bilan, Ivan Bogeski, Thomas Michel, Sophie Vriz, Joris Messens, Vsevolod V Belousov
ABSTRACT
Hydrogen peroxide (H2O2) is a key redox intermediate generated within cells. Existing probes for H2O2 have not solved the problem of detection of the ultra-low concentrations of the oxidant: these reporters are not sensitive enough, or pH-dependent, or insufficiently bright, or not functional in mammalian cells, or have poor dynamic range. Here we present HyPer7, the first bright, pH-stable, ultrafast, and ultrasensitive ratiometric H2O2 probe. HyPer7 is fully functional in mammalian cells and in other higher eukaryotes. The probe consists of a circularly permuted GFP integrated into the ultrasensitive OxyR domain from Neisseria meningitidis. Using HyPer7, we were able to uncover the details of H2O2 diffusion from the mitochondrial matrix, to find a functional output of H2O2 gradients in polarized cells, and to prove the existence of H2O2 gradients in wounded tissue in vivo. Overall, HyPer7 is a probe of choice for real-time H2O2 imaging in various biological contexts.
Sigma-Aldrich
Auranofin, ≥98% (HPLC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Xanthine Oxidase from bovine milk, lyophilized powder, 0.4-1.0 units/mg protein
Sigma-Aldrich
Cumene hydroperoxide, technical grade, 80%
Sigma-Aldrich
Benzamidine hydrochloride hydrate, ≥99%
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein