Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

104329

Sigma-Aldrich

2,3,4,5-Tetrafluoroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6F4NH2
CAS Number:
Molecular Weight:
165.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

bp

78 °C/13 mmHg (lit.)

mp

27-29 °C (lit.)

SMILES string

Nc1cc(F)c(F)c(F)c1F

InChI

1S/C6H3F4N/c7-2-1-3(11)5(9)6(10)4(2)8/h1H,11H2

InChI key

BEECAQIHCYTZHC-UHFFFAOYSA-N

General description

2,3,4,5-Tetrafluoroaniline reacts with glycerol, crotonaldehyde, and methyl vinyl ketone by Skraup reaction to form 5,6,7,8-Tetrafluoroquinoline and 2- and 4-methyl-5,6,7,8-tetrafluoroquinoline, respectively.

Application

2,3,4,5-Tetrafluoroaniline can be used in the synthesis of polyfluorinated quinolines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Partially fluorinated heterocyclic compounds. Part I. The preparation of some tetra-and tri-fluoroquinolines, and some reactions of 5, 6, 7, 8-tetrafluoroquinoline.
Brooke GM, et al.
J. Chem. Soc. Sect. C, 215-218 (1966)
N-Acetylation as a Means to Activate Polyfluoroarylamines for Selective ortho-Hydrodefluorination by Zinc in Aqueous Ammonia: A Concise Route to Polyfluorobenzo Azaheterocycles.
Laev SS, et al.
European Journal of Organic Chemistry, 2007(2), 306-316 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service