Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

102415

Sigma-Aldrich

1,2,3,4-Tetrahydronaphthalene

reagent grade, ≥97%

Synonym(s):

Tetralin solvent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
Beilstein:
1446407
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

reagent grade

vapor density

4.55 (vs air)

vapor pressure

0.18 mmHg ( 20 °C)

Assay

≥97%

autoignition temp.

723 °F

expl. lim.

0.8 %, 100 °F
5 %, 150 °F

refractive index

n20/D 1.541 (lit.)

bp

207 °C (lit.)

mp

−35 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

C1CCc2ccccc2C1

InChI

1S/C10H12/c1-2-6-10-8-4-3-7-9(10)5-1/h1-2,5-6H,3-4,7-8H2

InChI key

CXWXQJXEFPUFDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Tomás-Gallardo et al.
Applied and environmental microbiology, 78(15), 5460-5462 (2012-05-29)
Glucose catabolite repression of tetralin catabolic genes in Rhodococcus sp. strain TFB was shown to be exerted by a protein homologous to transcriptional regulators of the cyclic AMP receptor (CRP)-FNR family. The protein was detected bound to putative CRP-like boxes
Zere Konuspayeva et al.
Physical chemistry chemical physics : PCCP, 17(42), 28112-28120 (2015-03-15)
Au, Rh, Pd, Au-Rh and Au-Pd nanoparticles (NPs) were synthesized by colloidal chemical reduction and immobilized on hydrothermally-prepared rutile titania nanorods. The catalysts were characterized by aberration-corrected TEM/STEM, XPS, and FTIR, and were evaluated in the hydrogenation of tetralin in
Bin Ma et al.
Bioorganic & medicinal chemistry letters, 20(7), 2264-2269 (2010-03-02)
Modifying FTY720, an immunosuppressant modulator, led to a new series of well phosphorylated tetralin analogs as potent S1P1 receptor agonists. The stereochemistry effect of tetralin ring was probed, and (-)-(R)-2-amino-2-((S)-6-octyl-1,2,3,4-tetrahydronaphthalen-2-yl)propan-1-ol was identified as a good SphK2 substrate and potent S1P1
Jordi Casanovas et al.
Bioorganic chemistry, 33(6), 484-492 (2005-11-18)
The conformational properties of polygamain and morelensin, two aryltetralin lignan lactones, have been investigated in both the gas-phase and chloroform solution using DFT calculations at the B3LYP/6-311G(d,p) level. Results indicate that the conformation of polygamain is very rigid. Thus, the
Laura Tomás-Gallardo et al.
Microbial biotechnology, 2(2), 262-273 (2009-03-01)
The tetralin biodegradation pathway in Rhodococcus sp. strain TFB, a Gram-positive bacterium resistant to genetic manipulation, was characterized using a proteomic approach. Relative protein expression in cell free extracts from tetralin- and glucose-grown cells was compared using the 2D-DIGE technique.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service