MilliporeSigma
  • Involvement of a putative cyclic amp receptor protein (CRP)-like binding sequence and a CRP-like protein in glucose-mediated catabolite repression of thn genes in Rhodococcus sp. strain TFB.
Involvement of a putative cyclic amp receptor protein (CRP)-like binding sequence and a CRP-like protein in glucose-mediated catabolite repression of thn genes in Rhodococcus sp. strain TFB.
Applied and environmental microbiology (2012-05-29)
Laura Tomás-Gallardo, Eduardo Santero, Belén Floriano
ABSTRACT
Glucose catabolite repression of tetralin catabolic genes in Rhodococcus sp. strain TFB was shown to be exerted by a protein homologous to transcriptional regulators of the cyclic AMP receptor (CRP)-FNR family. The protein was detected bound to putative CRP-like boxes localized at the promoters of the thnA1 and thnS genes.
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, reagent grade, ≥97%
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, anhydrous, 99%
Sigma-Aldrich
1,2,3,4-Tetrahydronaphthalene, ReagentPlus®, 99%
Supelco
1,2,3,4-Tetrahydronaphthalene, analytical standard