Skip to Content
Merck
All Photos(1)

Key Documents

Clone M-3

87081806, mouse skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
mouse skin
growth mode:
Adherent
karyotype:
Hypertetraploid
morphology:
Epithelial
products:
Melanin
receptors:
Not specified
relevant disease(s):
cancer

Product Name

Clone M-3, 87081806

biological source

mouse skin

growth mode

Adherent

karyotype

Hypertetraploid

morphology

Epithelial

products

Melanin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse (CxDBA)F1 melanoma

Cell Line Description

Established from a Cloudman S91 melanoma in a male mouse; the cells produce melanin.

Application

Cloning studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 10% Horse Serum (HS) + 5% Foetal Bovine Serum (FBS) or 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service