The isoelectric point of the protein is pH 9.33, and its net charge at pH 7 is 25.9.
CAS9GFPPRO
Cas9-GFP Protein
from Streptococcus pyogenes, fused with enhanced GFP, recombinant, expressed in E. coli, 3X NLS
Synonym(s):
Cas9-EGFP, SpCas9-EGFP, SpCas9-GFP
About This Item
Recommended Products
recombinant
expressed in E. coli
Assay
≥90% (SDS-PAGE)
form
lyophilized powder
packaging
pkg of 1 kit (3 components)
reporter gene
GFP
shipped in
wet ice
storage temp.
−20°C
General description
An N-terminally fused enhanced green fluorescent protein with an excitation peak at 488 nm and emission peak at 509 nm allows for visualization of transfected RNP complex in addition to utility in flow cytometry applications. The protein also contains three varied nuclear localization sequences postioned for optimal activity.
Application
- Functional Genomics
- Target Validation
- Genome Editing
- Fluorescence Microscopy
- Flow Cytometry
Features and Benefits
- Highly specific
- Highly active
- Ready-to-inject/transfect
Packaging
- pkg of 50 μg( ≥260 pmol )
- pkg of 250 μg( ≥1300 pmol )
Components
- one vial of lyophilzed Cas9-GFP recombinant protein
- one vial containing 1 mL of 1× dilution buffer
- one vial containing 1 mL of nuclease-free water with glycerol
Principle
Reconstitution
Other Notes
Check out our other MISSION® Cas9 Proteins at SigmaAldrich.com/CRISPRproteins
Legal Information
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity
Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity
Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity
Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity
Related Content
Combine guaranteed sgRNAs with our comprehensive range of CRISPR products and tools, including Cas9 and delivery reagents, for efficient genome modification with higher specificity.
-
What are the net charge and isoelectric point of this protein?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service