Skip to Content
Merck
All Photos(1)

Key Documents

BCR576

Human serum (17β-estradiol, low level)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

clinical testing

format

matrix material

storage temp.

−20°C

Analysis Note

For more information please see:
BCR576

Other Notes

Sales restrictions may apply

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service