Skip to Content
Merck
All Photos(1)

Key Documents

MAB1407P

Sigma-Aldrich

Anti-Macrophages/Granulocytes Antibody, clone OX-41

clone OX-41, Chemicon®, from mouse

Synonym(s):

CD172 antigen-like family member A, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MyD-1 antigen, SHP substrate 1, SHP substrate-1, Signal-regulatory protein alpha-1, signal-regulatory protein alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

OX-41, monoclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SIRPA(140885)

Related Categories

General description

Macrophages and granulocytes express Signal regulatory protein (SIRP), which is a transmembrane glycoprotein involved in the down regulation of tyrosine kinase receptor signal transduction induced by cell adhesion, growth factors or insulin. They are immunoglobulin-like cell surface receptor for CD47. By CD47 binding there is a negative regulation of phagocytosis, mast cell activation and dendritic cell activation that prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells.

Specificity

Expected to cross-react with rat and mouse.
MAB1407P recognizes rat SIRP (Signal regulatory protein), which is expressed selectively by myeloid cells and by neurons. This monoclonal binds to a different epitope on SIRP than ED9. It does not block the interaction of CD172a and CD47.
Labels most macrophages including resident peritoneal, alveolar and activated macrophages. Recognition of other macrophages is limited to only occasional kupffer cells. In addition it labels granulocytes and a population of dendritic cells. It also labels brain extensively.

Immunogen

Rat peritoneal macrophages.

Application

Immunohistochemistry:
Frozen sections and Formalin fixed, paraffin embedded tissue sections (requires trypsin or pronase pre-treatment).

Western Blotting:
A previous lot of this antibody was used in western blot.

Flow cytometry:
1:100-1:200 of a previous lot was used. (Use 10 μL of working dilution for 10^6 cells in 100 μL).

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
This Anti-Macrophages/Granulocytes Antibody, clone OX-41 is validated for use in FC, IC, IH, IH(P), WB for the detection of Macrophages/Granulocytes.

Quality

Routinely evaluated by Immunocytochemistry on Macrophages/Granulocytes.

Immunocytochemistry Analysis:
Confolcal fluorescent analysis of Macrophages/ Granulocytes using anti-Macrophages/ Granulocytes mouse monoclonal antibody.

Target description

56 kDa

Linkage

Replaces: MAB1407

Physical form

ImmunoAffinity Purified
Purified mouse monoclonal IgG2a in buffer containing 0.09% sodium azide.

Storage and Stability

Stable for 6 months at 2-8°C in undiluted aliquots from date of receipt.

Analysis Note

Control
Macrophages, monocytes, lymphoid organs.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony Tam et al.
Respiratory research, 21(1), 46-46 (2020-02-09)
Female smokers have increased risk for chronic obstructive pulmonary disease (COPD) compared with male smokers who have a similar history of cigarette smoke exposure. Tertiary lymphoid follicles are often found in the lungs of patients with severe COPD but sex-related
Mao Nakayama et al.
Molecular vision, 17, 1784-1793 (2011-07-15)
This study was conducted to investigate the effect of flavonoids, a major family of antioxidants contained in foods, on retinal ganglion cell (RGC) death induced by hypoxia, excessive glutamate levels, and oxidative stress. Moreover, to assess the structure-activity relationships of
Xin-Mei Zhang et al.
Molecular vision, 16, 2867-2872 (2011-01-05)
To establish a robust method to isolate primary retinal ganglion cells (RGCs) from human fetal retina for long-term culture while maintaining neuronal morphology and marker protein expression. A total of six human retinas were obtained from aborted fetuses at 10
Xin Ding et al.
Nature communications, 12(1), 2030-2030 (2021-04-03)
Microglia play a key role in regulating synaptic remodeling in the central nervous system. Activation of classical complement pathway promotes microglia-mediated synaptic pruning during development and disease. CD47 protects synapses from excessive pruning during development, implicating microglial SIRPα, a CD47
Jun Nagai et al.
Scientific reports, 5, 8269-8269 (2015-02-06)
Axonal outgrowth inhibitors and scar formation are two major obstacles to central nervous system (CNS) repair. No target molecule that regulates both axonal growth and scarring has been identified. Here we identified collapsin response mediator protein 4 (CRMP4), a common

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service