Skip to Content
Merck
All Photos(1)

Key Documents

8.43827

Sigma-Aldrich

Diethylene glycol methyl ethyl ether

for synthesis

Synonym(s):

Diethylene glycol methyl ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16O3
CAS Number:
Molecular Weight:
148.20
UNSPSC Code:
12352112
EC Index Number:
213-690-5
NACRES:
NA.22

Quality Level

Assay

≥96.0% (GC)

form

liquid

potency

6000 mg/kg LD50, oral (Rat)
6500 mg/kg LD50, skin (Rabbit)

mp

-72 °C

transition temp

flash point 82 °C

solubility

76 g/L

density

0.925 g/cm3

storage temp.

2-30°C

InChI

1S/C7H16O3/c1-3-9-6-7-10-5-4-8-2/h3-7H2,1-2H3

InChI key

CNJRPYFBORAQAU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.923 - 0.927
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service