8.01313
2-Amino-2-thiazoline
for synthesis
Synonym(s):
2-Amino-2-thiazoline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H6N2S
CAS Number:
Molecular Weight:
102.16
MDL number:
UNSPSC Code:
12352116
EC Index Number:
217-224-1
NACRES:
NA.22
Recommended Products
Quality Level
form
crystals
mp
76-80 °C
storage temp.
2-30°C
SMILES string
S1CCNC1=N
InChI
1S/C3H6N2S/c4-3-5-1-2-6-3/h1-2H2,(H2,4,5)
InChI key
REGFWZVTTFGQOJ-UHFFFAOYSA-N
Analysis Note
Assay (HClO₄): ≥ 97.0 %
Melting range (lower value): ≥ 76 °C
Melting range (upper value): ≤ 80 °C
Identity (IR): passes test
Melting range (lower value): ≥ 76 °C
Melting range (upper value): ≤ 80 °C
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service