Skip to Content
Merck
All Photos(1)

Key Documents

218775

Sigma-Aldrich

Castanospermine, Castanospermum australe

Plant alkaloid inhibitor of several β-glucosidases and α-glucosidases, including those involved in N-linked processing of glycoproteins.

Synonym(s):

Castanospermine, Castanospermum australe, (1S,6S,7R,8aR)-Tetrahydroxyoctahydroindolizine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO4
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

description

Merck USA index - 14, 1896

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Plant alkaloid inhibitor of several β-glucosidases and α-glucosidases, including those involved in N-linked processing of glycoproteins. Resulting oligosaccharides are primarily Glu3Man7-9(GlcNAc)2. Reduces bFGF induced angiogenesis in mice. Inhibits endothelial cell migration and invasion of basement membrane. Exhibits anti-viral properties. Typically used at 1-10 µg/ml.

Biochem/physiol Actions

Cell permeable: no
Primary Target
β-glucosidases
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ahmed, S.P., et al. 1995. Biochem. Biophys. Res. Commun. 208, 267.
Pili, R., et al. 1995. Cancer Res.55, 2920.
Walker, B.D., et al. 1987. Proc. Natl. Acad. Sci. USA84, 8120.
Pan, Y.T., et al. 1983. Biochemistry22, 3975.
Saul, R., et al. 1983. Arch. Biochem. Biophys. 221, 593.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service