Key Documents
BMSIPD960010TCB
Sigma-Aldrich
BeadBlaster 96 Tungsten Carbide grinding ball
bead diam. 10 mm, Tungsten carbide grinding jar
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
material
tungsten carbide
packaging
pack of 2
manufacturer/tradename
Benchmark Scientific IPD9600-10TCB
bead diam.
10 mm
compatibility
Tungsten carbide grinding jar
General description
Tungsten carbide grinding media balls are the highest density media material for milling and crushing applications.
Tungsten carbide grinding media balls owns excellent performance in wear-resistance and resistance to acid and alkali. High hardness can meet the most majority of metal powder crushing and refining.
Application
Homogenization of metallurgy, geology and pharmaceutical samples
Features and Benefits
  • High density
  • Good resistance to acid and alkali
  • High hardness for wear resistance
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.